ENA066595634 Enamine

4-[(4-bromophenoxy)methyl]aniline

  • MDL number MFCD08271771
  • Cas Number : 885524-89-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15076

Safety Information

msds https://www.enaminestore.com/catalog/EN300-15076
Availability details for
Quantity
  • Availability
  • Add to Cart