ENA066595641 Enamine

5-ethoxy-2-methyl-2H,3H,8H-furo[3,2-g]indole-7-carboxylic acid

  • MDL number MFCD08262011
  • Cas Number : 879151-90-3
  • Molecular Weight : 261.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-15115

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-15115
Availability details for
Quantity
  • Availability
  • Add to Cart