ENA066595651 Enamine

N-[5-(azepan-1-ylsulfonyl)-2-methoxyphenyl]-2-chloroacetamide

  • MDL number MFCD07686046
  • Cas Number : 872107-76-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105...107
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15133

Safety Information

msds https://www.enaminestore.com/catalog/EN300-15133
Availability details for
Quantity
  • Availability
  • Add to Cart