ENA066595655 Enamine

1-(propan-2-yl)-1H-pyrazolo[3,4-b]pyridin-5-amine

  • MDL number MFCD07685830
  • Cas Number : 872103-27-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 85-86
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-15147

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-15147
Availability details for
Quantity
  • Availability
  • Add to Cart