ENA066595656 Enamine

1-(2,4-dichlorobenzyl)-1H-pyrazol-5-amine

  • MDL number MFCD07686041
  • Cas Number : 929028-88-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15151

Safety Information

msds https://www.enaminestore.com/catalog/EN300-15151
Availability details for
Quantity
  • Availability
  • Add to Cart