ENA066595662 Enamine

2,3-dimethyl-5-nitrobenzenesulfonyl chloride

  • MDL number MFCD08271784
  • Cas Number : 68631-04-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 103...105
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15175

Safety Information

msds https://www.enaminestore.com/catalog/EN300-15175
Availability details for
Quantity
  • Availability
  • Add to Cart