ENA066595669 Enamine

2-[(2-amino-2-oxoethyl)thio]nicotinic acid

  • MDL number MFCD03168919
  • Cas Number : 852695-22-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 162...164
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15186

Safety Information

msds https://www.enaminestore.com/catalog/EN300-15186
Availability details for
Quantity
  • Availability
  • Add to Cart