ENA066595672 Enamine

3-(2-oxo-5-thien-2-yl-1,3,4-oxadiazol-3(2H)-yl)propanoic acid

  • MDL number MFCD07686060
  • Cas Number : 872107-94-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 146...148
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15192

Safety Information

msds https://www.enaminestore.com/catalog/EN300-15192
Availability details for
Quantity
  • Availability
  • Add to Cart