ENA066595684 Enamine

2-(3-ethyl-2,4-dioxo-1,2,3,4-tetrahydroquinazolin-1-yl)acetic acid

  • MDL number MFCD07686069
  • Cas Number : 872108-02-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15216

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-15216
Availability details for
Quantity
  • Availability
  • Add to Cart