ENA066595686 Enamine

1-cyclopropyl-1H-1,2,3,4-tetrazole-5-thiol

  • MDL number MFCD08271790
  • Cas Number : 637781-10-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15219

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P310,P312,P330,P363,P501
MSDS https://www.enaminestore.com/catalog/EN300-15219
pictograms GHS05,GHS07
hstatements H302,H312,H318,H332
Availability details for
Quantity
  • Availability
  • Add to Cart