ENA066595689 Enamine

3-({imidazo[1,2-a]pyridin-2-yl}methoxy)benzoic acid

  • MDL number MFCD07686073
  • Cas Number : 872108-08-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15222

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-15222
Availability details for
Quantity
  • Availability
  • Add to Cart