ENA066595696 Enamine

N-{2-[4-(2-chloropropanoyl)phenyl]ethyl}acetamide

  • MDL number MFCD07687076
  • Cas Number : 105842-89-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 86...88
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15232

Safety Information

msds https://www.enaminestore.com/catalog/EN300-15232
Availability details for
Quantity
  • Availability
  • Add to Cart