ENA066595702 Enamine

methyl 2-(3,5-dimethyl-1H-pyrazol-4-yl)acetate

  • MDL number MFCD07685912
  • Cas Number : 56699-23-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 42-43
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15247

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-15247
Availability details for
Quantity
  • Availability
  • Add to Cart