ENA066595710 Enamine

3,4-dimethyl-5-nitrobenzenesulfonamide

  • MDL number MFCD07687329
  • Cas Number : 838838-15-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180...182
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15260

Safety Information

msds https://www.enaminestore.com/catalog/EN300-15260
Availability details for
Quantity
  • Availability
  • Add to Cart