ENA066595712 Enamine

3-[(2-fluorophenyl)sulfanyl]propanoic acid

  • MDL number MFCD06208520
  • Cas Number : 21243-09-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118-119
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15264

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-15264
Availability details for
Quantity
  • Availability
  • Add to Cart