ENA066595725 Enamine

N-{1-[4-(2-chloroacetyl)phenyl]ethyl}acetamide

  • MDL number MFCD07687989
  • Cas Number : 874594-00-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149-150
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-15293

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-15293
Availability details for
Quantity
  • Availability
  • Add to Cart