ENA066595735 Enamine

ethyl {3,5-dimethyl-1-[3-(trifluoromethyl)phenyl]-1H-pyrazol-4-yl}(oxo)acetate

  • MDL number MFCD07690491
  • Cas Number : 956438-46-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 50...52
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15311

Safety Information

msds https://www.enaminestore.com/catalog/EN300-15311
Availability details for
Quantity
  • Availability
  • Add to Cart