ENA066595746 Enamine

ethyl 2-(4-benzyl-5-sulfanyl-4H-1,2,4-triazol-3-yl)acetate

  • MDL number MFCD08271798
  • Cas Number : 874754-19-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-15334

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-15334
Availability details for
Quantity
  • Availability
  • Add to Cart