ENA066595749 Enamine

6-(2,3-dihydro-1,4-benzodioxin-6-yl)-3-thioxo-3,4-dihydro-1,2,4-triazin-5(2H)-one

  • MDL number MFCD07689454
  • Cas Number : 874754-24-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 190...192
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15339

Safety Information

msds https://www.enaminestore.com/catalog/EN300-15339
Availability details for
Quantity
  • Availability
  • Add to Cart