ENA066595750 Enamine

6-(2,5-dichlorothien-3-yl)-3-thioxo-3,4-dihydro-1,2,4-triazin-5(2H)-one

  • MDL number MFCD07689455
  • Cas Number : 874754-25-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 203...205
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15340

Safety Information

msds https://www.enaminestore.com/catalog/EN300-15340
Availability details for
Quantity
  • Availability
  • Add to Cart