ENA066595753 Enamine

3-(2-oxo-3-propyl-2,3-dihydro-1H-benzimidazol-1-yl)propanoic acid

  • MDL number MFCD07689458
  • Cas Number : 874754-26-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 127...129
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15343

Safety Information

msds https://www.enaminestore.com/catalog/EN300-15343
Availability details for
Quantity
  • Availability
  • Add to Cart