ENA066595771 Enamine

cyclohexanecarbonyl isothiocyanate

  • MDL number MFCD08271814
  • Cas Number : 27699-51-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-21044

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-21044
Availability details for
Quantity
  • Availability
  • Add to Cart