ENA066595772 Enamine

5-phenyl-3-(propan-2-yl)-2-sulfanyl-3H,4H-thieno[2,3-d]pyrimidin-4-one

  • MDL number MFCD08271815
  • Cas Number : 885525-67-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-21607

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-21607
Availability details for
Quantity
  • Availability
  • Add to Cart