ENA066595778 Enamine

3-[2-oxo-6-(pyrrolidine-1-sulfonyl)-2,3-dihydro-1,3-benzoxazol-3-yl]propanoic acid

  • MDL number MFCD07687082
  • Cas Number : 873790-30-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 190-191
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-21797

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-21797
Availability details for
Quantity
  • Availability
  • Add to Cart