ENA066595796 Enamine

4-[(2-chlorophenyl)amino]quinazoline-2-thiol

  • MDL number MFCD08271821
  • Cas Number : 873790-39-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-21833

Safety Information

msds https://www.enaminestore.com/catalog/EN300-21833
Availability details for
Quantity
  • Availability
  • Add to Cart