ENA066595797 Enamine

4-[5-(4-methylphenyl)-1,3,4-oxadiazol-2-yl]benzoic acid

  • MDL number MFCD07687275
  • Cas Number : 873208-67-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 250-251
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-21834

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-21834
Availability details for
Quantity
  • Availability
  • Add to Cart