ENA066595798 Enamine

3-(3-cyclopropyl-4-oxo-3,4-dihydroquinazolin-2-yl)propanoic acid

  • MDL number MFCD07687286
  • Cas Number : 873208-74-3
  • Molecular Weight : 258.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-21838

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-21838
Availability details for
Quantity
  • Availability
  • Add to Cart