ENA066595803 Enamine

2-chloro-N-ethyl-N-({[(thiophen-2-yl)methyl]carbamoyl}methyl)acetamide

  • MDL number MFCD07690493
  • Cas Number : 874594-85-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-21858

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-21858
Availability details for
Quantity
  • Availability
  • Add to Cart