ENA066595804 Enamine

N-butyl-N-(sec-butyl)-2-chloroacetamide

  • MDL number MFCD07690494
  • Cas Number : 874594-86-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-21859

Safety Information

msds https://www.enaminestore.com/catalog/EN300-21859
Availability details for
Quantity
  • Availability
  • Add to Cart