ENA066595805 Enamine

2-chloro-N-cyclopropyl-N-[(4-methoxyphenyl)methyl]acetamide

  • MDL number MFCD07690495
  • Cas Number : 874594-87-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-21860

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-21860
Availability details for
Quantity
  • Availability
  • Add to Cart