ENA066595807 Enamine

2-chloro-N-cyclopropyl-N-[(2-fluorophenyl)methyl]acetamide

  • MDL number MFCD07690515
  • Cas Number : 874595-08-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-21862

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-21862
Availability details for
Quantity
  • Availability
  • Add to Cart