ENA066595817 Enamine

6-amino-1-ethyl-5-[2-(4-hydroxyphenyl)-1,3-thiazol-4-yl]pyrimidine-2,4(1H,3H)-dione

  • MDL number MFCD07352320
  • Cas Number : 871544-59-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15016

Safety Information

msds https://www.enaminestore.com/catalog/EN300-15016
Availability details for
Quantity
  • Availability
  • Add to Cart