ENA066595823 Enamine

2-[(carbamoylmethyl)sulfanyl]benzoic acid

  • MDL number MFCD00031447
  • Cas Number : 197236-53-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210-211
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15180

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-15180
Availability details for
Quantity
  • Availability
  • Add to Cart