ENA066595835 Enamine

3-methyl-5-(methylamino)isothiazole-4-carbonitrile

  • MDL number MFCD07366455
  • Cas Number : 88394-37-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140...142
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15027

Safety Information

msds https://www.enaminestore.com/catalog/EN300-15027
Availability details for
Quantity
  • Availability
  • Add to Cart