ENA066595838 Enamine

3-(2-chloroacetyl)-1-[(thiophen-2-yl)methyl]urea

  • MDL number MFCD07366456
  • Cas Number : 871909-81-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170-171
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-15029

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-15029
Availability details for
Quantity
  • Availability
  • Add to Cart