ENA066595841 Enamine

4-phenoxybutan-1-amine

  • MDL number MFCD00031400
  • Cas Number : 16728-66-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15198

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-15198
Availability details for
Quantity
  • Availability
  • Add to Cart