ENA066595852 Enamine

5-(furan-2-yl)-5-methylimidazolidine-2,4-dione

  • MDL number MFCD00461529
  • Cas Number : 4615-71-8
  • Molecular Weight : 180.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 174-177
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-15209

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-15209
Availability details for
Quantity
  • Availability
  • Add to Cart