ENA066595870 Enamine

3-(4-acetyl-5-methylfuran-2-yl)propanoic acid

  • MDL number MFCD07355838
  • Cas Number : 858247-32-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90-95
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15047

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-15047
Availability details for
Quantity
  • Availability
  • Add to Cart