ENA066595877 Enamine

1-[5-(methylamino)-1,2,4-thiadiazol-3-yl]propan-2-one

  • MDL number MFCD00173993
  • Cas Number : 257862-97-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15234

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-15234
s_phrases_eu 7-16-36/37-45
Availability details for
Quantity
  • Availability
  • Add to Cart