ENA066595884 Enamine

2-{[(2-methyl-1,3-thiazol-4-yl)methyl]sulfanyl}acetic acid

  • MDL number MFCD03011607
  • Cas Number : 446829-98-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-15241

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-15241
Availability details for
Quantity
  • Availability
  • Add to Cart