ENA066595889 Enamine

5-{[(4-methoxyphenyl)methyl]amino}-1,3,4-thiadiazole-2-thiol

  • MDL number MFCD08271793
  • Cas Number : 873790-27-3
  • Molecular Weight : 253.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 137-138
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15246

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-15246
Availability details for
Quantity
  • Availability
  • Add to Cart