ENA066595911 Enamine

1-acetyl-1,2,3,4-tetrahydroquinoline-6-sulfonyl chloride

  • MDL number MFCD07366466
  • Cas Number : 868964-04-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90-91
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15069

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-15069
Availability details for
Quantity
  • Availability
  • Add to Cart