ENA066595917 Enamine

5-{[1,1'-biphenyl]-4-yl}-1-(4-methylphenyl)-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD02942159
  • Cas Number : 955972-89-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15072

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-15072
Availability details for
Quantity
  • Availability
  • Add to Cart