ENA066595921 Enamine

4-[(2-isopropyl-5-methylphenoxy)methyl]aniline

  • MDL number MFCD02063331
  • Cas Number : 329063-78-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15073

Safety Information

msds https://www.enaminestore.com/catalog/EN300-15073
Availability details for
Quantity
  • Availability
  • Add to Cart