ENA066595931 Enamine

2-chloro-4H-pyrido[1,2-a]pyrimidin-4-one

  • MDL number MFCD00457859
  • Cas Number : 5418-94-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155-160
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15294

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-15294
Availability details for
Quantity
  • Availability
  • Add to Cart