ENA066595935 Enamine

6-chloro-1H-indole-2-carboxylic acid

  • MDL number MFCD01863157
  • Cas Number : 16732-75-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 242...244
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15085

Safety Information

msds https://www.enaminestore.com/catalog/EN300-15085
Availability details for
Quantity
  • Availability
  • Add to Cart