ENA066595941 Enamine

2-[carbamoyl(methyl)amino]acetic acid

  • MDL number MFCD07688008
  • Cas Number : 30565-25-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142-143
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-15305

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-15305
Availability details for
Quantity
  • Availability
  • Add to Cart