ENA066595957 Enamine

4-ethoxy-1H-indole-2-carboxylic acid

  • MDL number MFCD02664459
  • Cas Number : 29970-01-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 255...257
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15097

Safety Information

msds https://www.enaminestore.com/catalog/EN300-15097
Availability details for
Quantity
  • Availability
  • Add to Cart