ENA066595963 Enamine

6-(benzyloxy)-1H-indole-2-carboxylic acid

  • MDL number MFCD02664461
  • Cas Number : 40047-22-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15100

Safety Information

msds https://www.enaminestore.com/catalog/EN300-15100
Availability details for
Quantity
  • Availability
  • Add to Cart