ENA066595968 Enamine

6-methyl-1H-indole-2-carboxylic acid

  • MDL number MFCD02664477
  • Cas Number : 18474-59-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-15104

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-15104
Availability details for
Quantity
  • Availability
  • Add to Cart